accessibility menu, dialog, popup

UserName

CAS RN 852181-17-0

CH3OHOF

CAS RN 852181-17-0

IUPAC Name: 3-(2-fluoro-4-methylphenyl)propanoic acid
Synonyms: 3-(2-fluoro-4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H11FO2
InChi Key: HQOAGAGIHCHXLP-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(CCC(O)=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results