CAS RN 852403-68-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(E)-Metaflumizone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
(E)-Metaflumizone, TRC
CAS: 852403-68-0 Molecular Formula: C24 H16 F6 N4 O2 Molecular Weight (g/mol): 506.4 Synonym: (2E)-2-[2-(4-Cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]-N-[4-(trifluoromethoxy)phenyl]hydrazinecarboxamide,(E)-Metaflumizone IUPAC Name: 1-[(E)-[2-(4-cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]amino]-3-[4-(trifluoromethoxy)phenyl]urea SMILES: FC(F)(F)Oc1ccc(NC(=O)N\N=C(/Cc2ccc(cc2)C#N)\c3cccc(c3)C(F)(F)F)cc1