CAS RN 85559-61-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-O-Alpha-D-Glucosyl Isomaltol, TRC
CAS: 85559-61-1 Molecular Formula: C12H16O8 Molecular Weight (g/mol): 288.25 Synonym: 1-[3-(α-D-Glucopyranosyloxy)-2-furanyl]ethanone,2-Acetylfuran-3yl-α-D-glucopyranoside; IUPAC Name: 1-[3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyfuran-2-yl]ethanone SMILES: O=C(C)C(OC=C1)=C1O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2