accessibility menu, dialog, popup

UserName

CAS RN 85952-73-4

NHF

CAS RN 85952-73-4

IUPAC Name: N-[(4-fluorophenyl)methyl]cyclopentanamine
Synonyms: N-[(4-fluorophenyl)methyl]cyclopentanamine
Molecular Weight (g/mol): 193.27
Molecular Formula: C12H16FN
InChi Key: AZUJIJXHVNXASI-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results