accessibility menu, dialog, popup

UserName

CH3OOSCl

CAS RN 85992-30-9

IUPAC Name: methyl 1-chloronaphtho[2,1-b]thiophene-2-carboxylate
Synonyms: methyl 1-chloronaphtho[2,1-b]thiophene-2-carboxylate
Molecular Weight (g/mol): 276.73
Molecular Formula: C14H9ClO2S
InChi Key: OHOGUWDSHRBKSE-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C2=C(S1)C=CC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results