accessibility menu, dialog, popup

UserName

CAS RN 86120-40-3

IUPAC Name: methyl (2E)-4-(dimethoxyphosphoryl)but-2-enoate
Synonyms: methyl (2E)-4-(dimethoxyphosphoryl)but-2-enoate
Molecular Weight (g/mol): 208.15
Molecular Formula: C7H13O5P
InChi Key: PESRINKNQQZURC-SNAWJCMRSA-N
SMILES: COC(=O)\C=C\CP(=O)(OC)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results