CAS RN 86129-54-6
CAS RN 86129-54-6
IUPAC Name:
6-chloro-3-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride
Synonyms:
benalfocin hydrochloride
Molecular Weight (g/mol):
232.15
Molecular Formula:
C11H15Cl2N
InChi Key:
JKQKVNMNAIFCFS-UHFFFAOYSA-N
SMILES:
Cl.CN1CCC2=C(CC1)C(Cl)=CC=C2
1
–
1
of
1
results
1
Tocris Bioscience™ SKF 86466 hydrochloride
CAS: 86129-54-6 Molecular Formula: C11H15Cl2N