accessibility menu, dialog, popup

UserName

CAS RN 86393-32-0

HH2OClOHONNNHFO

CAS RN 86393-32-0

IUPAC Name: hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
Synonyms: hydrogen ciprofloxacin hydrate chloride
Molecular Weight (g/mol): 385.82
Molecular Formula: C17H21ClFN3O4
InChi Key: ARPUHYJMCVWYCZ-UHFFFAOYSA-N
SMILES: [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)