accessibility menu, dialog, popup

UserName

H3COPOOCH3ClClCl

CAS RN 866-23-9

IUPAC Name: diethyl (trichloromethyl)phosphonate
Synonyms: diethyl trichloromethylphosphonate
Molecular Weight (g/mol): 255.46
Molecular Formula: C5H10Cl3O3P
InChi Key: RVAQSYWDOSHWGP-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C(Cl)(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results