accessibility menu, dialog, popup

UserName

CAS RN 868755-53-7

ONNN

CAS RN 868755-53-7

IUPAC Name: 6-(morpholin-4-yl)pyridine-2-carbonitrile
Synonyms: 6-(morpholin-4-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: WKDAOTSKEPJLCC-UHFFFAOYSA-N
SMILES: N#CC1=NC(=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results