accessibility menu, dialog, popup

UserName

CAS RN 868755-61-7

H3COONNH3CS

CAS RN 868755-61-7

IUPAC Name: ethyl 1-methyl-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate
Synonyms: ethyl 2-methyl-5-(thiophen-2-yl)pyrazole-3-carboxylate
Molecular Weight (g/mol): 236.29
Molecular Formula: C11H12N2O2S
InChi Key: MUIUODSZPZJMMP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=NN1C)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results