CAS RN 868755-62-8
CAS RN 868755-62-8
IUPAC Name:
5-(furan-2-yl)thiophene-2-carboxylic acid
Synonyms:
5-(furan-2-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol):
194.2
Molecular Formula:
C9H6O3S
InChi Key:
ZASUJVQPFQRFPQ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(S1)C1=CC=CO1