CAS RN 86878-53-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Disodium Monooctyl Sulfosuccinate, TRC
CAS: 86878-53-7 Molecular Formula: C12H20Na2O7S Molecular Weight (g/mol): 354.33 Synonym: Docusate Sodium Impurity,1-(2-Ethylhexyl)ester-2-sulfo-butanedioic Acid Sodium Salt; IUPAC Name: disodium;4-(2-ethylhexoxy)-4-oxo-3-sulfonatobutanoate SMILES: O=C(C(S(=O)(O[Na])=O)CC(O[Na])=O)OCC(CCCC)CC