CAS RN 86900-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R,S)-N-Ethyl Nornicotine, TRC
CAS: 86900-39-2 Molecular Formula: C11 H16 N2 Molecular Weight (g/mol): 176.26 Synonym: Pyridine, 3-(1-ethyl-2-pyrrolidinyl)-,Pyridine, 3-(1-ethyl-2-pyrrolidinyl)-, (±)-,3-(1-Ethyl-2-pyrrolidinyl)pyridine,Homonicotine,(±)-N-Ethylnornicotine,rac-Ethylnornicotine,rac-N-Ethylnornicotine IUPAC Name: 3-(1-ethylpyrrolidin-2-yl)pyridine SMILES: CCN1CCCC1c2cccnc2