CAS RN 869111-60-4
CAS RN 869111-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(3S,3'S,5S,5'S,6R,6'R)3,3'-[[(Phenylmethyl)imino]bis[(3S)-3-hydroxy-4,1-butanediyl]]bis[2-oxo-5,6-diphenyl-4-morpholinecarboxylic Acid Bis(tert-butyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3S,3'S,5S,5'S,6R,6'R)3,3'-[[(Phenylmethyl)imino]bis[(3S)-3-hydroxy-4,1-butanediyl]]bis[2-oxo-5,6-diphenyl-4-morpholinecarboxylic Acid Bis(tert-butyl) Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.