CAS RN 869287-60-5
CAS RN 869287-60-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methandrostenolone-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methandrostenolone-d3 (1 mg/ml in Acetonitrile), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.