accessibility menu, dialog, popup

UserName

H3CSOONHNNFFFNHHNO

CAS RN 869288-64-2

IUPAC Name: 6-[(4-{[(3-methanesulfonylphenyl)methyl]amino}-5-(trifluoromethyl)pyrimidin-2-yl)amino]-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 6-[(4-{[(3-methanesulfonylphenyl)methyl]amino}-5-(trifluoromethyl)pyrimidin-2-yl)amino]-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol): 491.49
Molecular Formula: C22H20F3N5O3S
InChi Key: HESLKTSGTIBHJU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC(CNC2=NC(NC3=CC4=C(NC(=O)CC4)C=C3)=NC=C2C(F)(F)F)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results