accessibility menu, dialog, popup

UserName

NBrH2NF

CAS RN 869557-43-7

IUPAC Name: 3-bromo-5-fluoropyridin-2-amine
Synonyms: 3-bromo-5-fluoropyridin-2-amine
Molecular Weight (g/mol): 191
Molecular Formula: C5H4BrFN2
InChi Key: KXSQMCRVUAALNE-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results