CAS RN 869627-83-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Estradiol 17-Isovalerate, TRC
CAS: 869627-83-8 Molecular Formula: C23H32O3 Molecular Weight (g/mol): 356.51 Synonym: (17beta)-Estra-1,3,5(10)-triene-3,17-diol 17-(3-Methylbutanoate) SMILES: CC(C)CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C