accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOHO

CAS RN 869681-70-9

IUPAC Name: 4-[(tert-butoxy)carbonyl]morpholine-3-carboxylic acid
Synonyms: 4-(tert-butoxycarbonyl)morpholine-3-carboxylic acid
Molecular Weight (g/mol): 231.25
Molecular Formula: C10H17NO5
InChi Key: KVXXEKIGMOEPSA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCOCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)