CAS RN 86978-24-7
CAS RN 86978-24-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(Z)-2-[[(1,1-Dimethylethoxy)carbonyl]amino]-Alpha-propylidene-4-thiazoleacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(Z)-2-[[(1,1-Dimethylethoxy)carbonyl]amino]-Alpha-propylidene-4-thiazoleacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.