accessibility menu, dialog, popup

UserName

CAS RN 869901-04-2

IUPAC Name: 2-(bromomethyl)-6-(pyrrolidin-1-yl)pyridine
Synonyms: 2-(bromomethyl)-6-(pyrrolidin-1-yl)pyridine
Molecular Weight (g/mol): 241.13
Molecular Formula: C10H13BrN2
InChi Key: UFXKRINLXMWRES-UHFFFAOYSA-N
SMILES: BrCC1=NC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results