CAS RN 869901-13-3
IUPAC Name:
3-(chloromethyl)-1-methyl-5-phenyl-1H-pyrazole
Synonyms:
3-(chloromethyl)-1-methyl-5-phenylpyrazole
Molecular Weight (g/mol):
206.67
Molecular Formula:
C11H11ClN2
InChi Key:
KAZKAWDJSYXNGK-UHFFFAOYSA-N
SMILES:
CN1N=C(CCl)C=C1C1=CC=CC=C1