CAS RN 869901-16-6
CAS RN 869901-16-6
IUPAC Name:
{[5-(furan-2-yl)thiophen-2-yl]methyl}(methyl)amine
Synonyms:
{[5-(furan-2-yl)thiophen-2-yl]methyl}(methyl)amine
Molecular Weight (g/mol):
193.26
Molecular Formula:
C10H11NOS
InChi Key:
REBAUXOPYFOJIE-UHFFFAOYSA-N
SMILES:
CNCC1=CC=C(S1)C1=CC=CO1