CAS RN 87-01-4
IUPAC Name:
7-(dimethylamino)-4-methyl-2H-chromen-2-one
Synonyms:
7-(dimethylamino)-4-methylchromen-2-one
Molecular Weight (g/mol):
203.24
Molecular Formula:
C12H13NO2
InChi Key:
GZEYLLPOQRZUDF-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C2C(C)=CC(=O)OC2=C1
1
–
3
of
3
results
1
7-(Dimethylamino)-4-methylcoumarin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-(Dimethylamino)-4-methylcoumarin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-(Dimethylamino)-4-methylcoumarin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.