CAS RN 87-02-5
CAS RN 87-02-5
IUPAC Name:
7-amino-4-hydroxynaphthalene-2-sulfonic acid
Synonyms:
7-amino-4-hydroxynaphthalene-2-sulfonic acid
Molecular Weight (g/mol):
239.25
Molecular Formula:
C10H9NO4S
InChi Key:
KYARBIJYVGJZLB-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C(O)=CC(=CC2=C1)S(O)(=O)=O
1
–
3
of
3
results
1
2-Amino-5-naphthol-7-sulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-5-naphthol-7-sulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Amino-5-naphthol-7-sulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.