accessibility menu, dialog, popup

UserName

OHONH

CAS RN 87-17-2

IUPAC Name: 2-hydroxy-N-phenylbenzamide
Synonyms: salicylanilide salinide salicylic acid anilide 2-N-phenylcarboxamidophenol
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: WKEDVNSFRWHDNR-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results