CAS RN 87-40-1
CAS RN 87-40-1
IUPAC Name:
1,3,5-trichloro-2-methoxybenzene
Synonyms:
tyrene C7H5cl3O
Molecular Weight (g/mol):
211.47
Molecular Formula:
C7H5Cl3O
InChi Key:
WCVOGSZTONGSQY-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C=C(Cl)C=C1Cl
1
–
2
of
2
results
1
2,4,6-Trichloroanisole 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality