CAS RN 87-83-2
CAS RN 87-83-2
IUPAC Name:
1,2,3,4,5-pentabromo-6-methylbenzene
Synonyms:
pentabromotoluene
Molecular Weight (g/mol):
486.62
Molecular Formula:
C7H3Br5
InChi Key:
OZHJEQVYCBTHJT-UHFFFAOYSA-N
SMILES:
CC1=C(Br)C(Br)=C(Br)C(Br)=C1Br