accessibility menu, dialog, popup

UserName

CAS RN 870-93-9

NH2SSNH2HOOOHO

CAS RN 870-93-9

IUPAC Name: 2-amino-4-[(3-amino-3-carboxypropyl)disulfanyl]butanoic acid
Synonyms: homocystine L-homocystine 4,4'-dithiobis DL-homocystine homocystine, L-
Molecular Weight (g/mol): 268.35
Molecular Formula: C8H16N2O4S2
InChi Key: ZTVZLYBCZNMWCF-UHFFFAOYNA-N
SMILES: NC(CCSSCCC(N)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)