CAS RN 870076-72-5
CAS RN 870076-72-5
IUPAC Name:
4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
Synonyms:
4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol
Molecular Weight (g/mol):
253.34
Molecular Formula:
C14H23NO3
InChi Key:
UMHASVFLCHGDPW-UHFFFAOYNA-N
SMILES:
COC(CNC(C)(C)C)C1=CC(CO)=C(O)C=C1
1
–
1
of
1
results
1
Alpha-Methoxy Albuterol, TRC
CAS: 870076-72-5 Molecular Formula: C14 H23 N O3 Molecular Weight (g/mol): 253.34 Synonym: Benzenemethanol, 5-[2-[(1,1-dimethylethyl)amino]-1-methoxyethyl]-2-hydroxy-,4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol,Salbutamol Sulphate Imp A (EP),[5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol IUPAC Name: 4-[2-(tert-butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol SMILES: COC(CNC(C)(C)C)c1ccc(O)c(CO)c1