CAS RN 87022-36-4
CAS RN 87022-36-4
IUPAC Name:
N-[(1H-1,2,3-benzotriazol-1-yl)methyl]formamide
Synonyms:
N-(1,2,3-benzotriazol-1-ylmethyl)formamide
Molecular Weight (g/mol):
176.18
Molecular Formula:
C8H8N4O
InChi Key:
AUDGAMZNNDGYAX-UHFFFAOYSA-N
SMILES:
O=CNCN1N=NC2=CC=CC=C12