accessibility menu, dialog, popup

UserName

CAS RN 870244-34-1

NBrNBr

CAS RN 870244-34-1

IUPAC Name: 3,5-dibromopyridine-4-carbonitrile
Synonyms: 3,5-dibromopyridine-4-carbonitrile
Molecular Weight (g/mol): 261.9
Molecular Formula: C6H2Br2N2
InChi Key: FHVATNRRDOWTPX-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(Br)=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results