CAS RN 870634-36-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 5-(4-Fluorophenyl)-(5S)-hydroxypentanoate, TRC
CAS: 870634-36-9 Molecular Formula: C12H15FO3 Molecular Weight (g/mol): 226.24 Synonym: Methyl Ester 4-Fluoro-delta-hydroxybenzenepentanoic Acid, IUPAC Name: methyl (5S)-5-(4-fluorophenyl)-5-hydroxypentanoate SMILES: COC(=O)CCC[C@H](O)c1ccc(F)cc1