accessibility menu, dialog, popup

UserName

CAS RN 870717-91-2

BOOH3CCH3H3CH3CCH3(E/Z)

CAS RN 870717-91-2

IUPAC Name: 2-[2-(4-ethylphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(4-ethylphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 258.17
Molecular Formula: C16H23BO2
InChi Key: WXXFAINZQODXKD-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results