accessibility menu, dialog, popup

UserName

CAS RN 870773-76-5

H3COOSNOOHFH3C

CAS RN 870773-76-5

IUPAC Name: 4-[4-(2-butoxyethoxy)-5-methyl-1,3-thiazol-2-yl]-2-fluorobenzoic acid
Synonyms: 4-[4-(2-butoxyethoxy)-5-methyl-1,3-thiazol-2-yl]-2-fluorobenzoic acid
Molecular Weight (g/mol): 353.41
Molecular Formula: C17H20FNO4S
InChi Key: HSAOETBFVAWNRP-UHFFFAOYSA-N
SMILES: CCCCOCCOC1=C(C)SC(=N1)C1=CC(F)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results