CAS RN 870823-12-4
CAS RN 870823-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MK 0893, MedChemExpress
MedChemExpress MK 0893 is a potent and selective glucagon receptor antagonist with an IC50 of 6.6 nM.