accessibility menu, dialog, popup

UserName

CAS RN 871-58-9

KH3COSS

CAS RN 871-58-9

IUPAC Name: potassium (butoxymethanethioyl)sulfanide
Synonyms: potassium (butoxymethanethioyl)sulfanide
Molecular Weight (g/mol): 188.34
Molecular Formula: C5H9KOS2
InChi Key: OMKVZYFAGQKILB-UHFFFAOYSA-M
SMILES: [K+].CCCCOC([S-])=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)