CAS RN 871-70-5
IUPAC Name:
octadecanedioic acid
Synonyms:
octadecanedioic acid
Molecular Weight (g/mol):
314.47
Molecular Formula:
C18H34O4
InChi Key:
BNJOQKFENDDGSC-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCCCCCCCCC(O)=O
1
–
2
of
2
results
1
Octadecanedioic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Octadecanedioic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.