accessibility menu, dialog, popup

UserName

HOCH

CAS RN 871-91-0

IUPAC Name: oct-7-yn-1-ol
Synonyms: oct-7-yn-1-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: ATCNYMVVGBLQMQ-UHFFFAOYSA-N
SMILES: OCCCCCCC#C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results