accessibility menu, dialog, popup

UserName

BFFFFFK

CAS RN 871231-41-3

IUPAC Name: potassium (2,4-difluorophenyl)trifluoroboranuide
Synonyms: potassium (2,4-difluorophenyl)trifluoroboranuide
Molecular Weight (g/mol): 219.99
Molecular Formula: C6H3BF5K
InChi Key: HZKKNPYRMYVEOW-UHFFFAOYSA-N
SMILES: [K+].FC1=CC(F)=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results