accessibility menu, dialog, popup

UserName

CAS RN 871252-64-1

IUPAC Name: 4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
Synonyms: 4-[5-(trifluoromethyl)pyridin-2-yl]benzaldehyde
Molecular Weight (g/mol): 251.21
Molecular Formula: C13H8F3NO
InChi Key: SBXWMOPSHGIWEN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results