CAS RN 871266-63-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PCI-27483, MedChemExpress
MedChemExpress PCI-27483 is a FVIIa/tissue factor inhibitor, with antitumour effects.