CAS RN 871329-30-5
CAS RN 871329-30-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-[(4-Hydroxyphenyl)methyl]oxolan-2-one, TRC
CAS: 871329-30-5 Molecular Formula: C11H12O3 Molecular Weight (g/mol): 192.21 IUPAC Name: 5-[(4-hydroxyphenyl)methyl]oxolan-2-one SMILES: Oc1ccc(CC2CCC(=O)O2)cc1