accessibility menu, dialog, popup

UserName

BSOONHHOOH

CAS RN 871329-67-8

IUPAC Name: [4-(cyclopropylsulfamoyl)phenyl]boronic acid
Synonyms: 4-(cyclopropylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 241.07
Molecular Formula: C9H12BNO4S
InChi Key: FUEGEHDCXRTJPN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)S(=O)(=O)NC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results