CAS RN 87152-97-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
trans-3,4-Dihydro-6-[4-[1-[4-(phenylmethoxy)cyclohexyl]-1H-tetrazol-5-yl]butoxy]-2(1H)-quinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans-3,4-Dihydro-6-[4-[1-[4-(phenylmethoxy)cyclohexyl]-1H-tetrazol-5-yl]butoxy]-2(1H)-quinolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.