CAS RN 87153-04-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4' '-trans-Hydroxy Cilostazol, TRC
CAS: 87153-04-6 Molecular Formula: C20 H27 N5 O3 Molecular Weight (g/mol): 385.46 Synonym: 4' '-Trans-Hydroxy Cilostazol IUPAC Name: 6-[4-[1-(4-hydroxycyclohexyl)tetrazol-5-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one SMILES: O[C@@H]1CC[C@H](CC1)n2nnnc2CCCCOc3ccc4NC(=O)CCc4c3