CAS RN 871550-18-4
CAS RN 871550-18-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[2,4-Bis(1,2,4-triazol-1-yl)phenyl]-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.