accessibility menu, dialog, popup

UserName

CAS RN 87179-40-6

NNH(E/Z)

CAS RN 87179-40-6

IUPAC Name: 1-(3-phenylprop-2-en-1-yl)piperazine
Synonyms: 1-(3-phenylprop-2-en-1-yl)piperazine
Molecular Weight (g/mol): 202.3
Molecular Formula: C13H18N2
InChi Key: WGEIOMTZIIOUMA-UHFFFAOYSA-N
SMILES: C(C=CC1=CC=CC=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results