CAS RN 871894-35-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 2-Chloro Nicotine, TRC
CAS: 871894-35-8 Molecular Formula: C10H13ClN2 Molecular Weight (g/mol): 196.68 Synonym: 2-Chloro-3-(tetrahydro-1-methyl-2-pyrryl)pyridine,2-Chloro-3-(1-methyl-2-pyrrolidinyl)pyridine IUPAC Name: 2-chloro-3-(1-methylpyrrolidin-2-yl)pyridine SMILES: Fc1cc2[C@@H]3[C@@H](CC=C3)C(Nc2c(F)c1)c4cnccc4